• USA Home
  • C4589 - EZMix Hy-Case® SF

EMAIL THIS PAGE TO A FRIEND
C4589 Sigma

EZMix Hy-Case® SF

Casein acid hydrolysate, from bovine milk

Synonym: Casein hydrolyzates

Purchase

Properties

biological source   from bovine milk
grade   for molecular biology
sterility   non-sterile
form   powder
storage temp.   2-8°C
suitability   nonselective for Escherichia coli and Coliforms

Description

Application

Recommended for use as a microbiological nutrient in laboratory media, fermentations, and tissue culture applications requiring a mixture of highly soluble free amino acids.

Features and Benefits

EZMix Hy-Case SF uses the same preparation instructions as Hy-Case SF. The EZMix formulation has the added advantage of being granulated and dust-free, allowing easier dissolution and safer and easier weighing and handling.

Other Notes

The EZMix powders provide the advantage of being granulated and dust-free. Therefore, because there is no dust hazard, safer and more accurate measurements can be taken. In addition, the EZmix powders dissolve faster and more completely than standard media. For more routine formulations, packet sizes have been pre-measured for added convenience.

Linkage

The growth characteristics are the same as Hy-Case SF.

Legal Information

EZMix is a trademark of Sigma-Aldrich Co. LLC

Hy-Case is a registered trademark of Kerry Group

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?