• USA Home
  • C4604 - Exposure cassette Carestream® Kodak® BioMax®
EMAIL THIS PAGE TO A FRIEND
C4604 Sigma
Exposure cassette Carestream® Kodak® BioMax®
For 14 in. x 17 in. (35 x 43 cm) film
Synonym: autoradiography cassettes, xray film cassettes
Purchase
Description
General description
Lightweight, durable, vinyl-covered aluminum cassettes, light-tight with single locking clip.
When used at ultra-cold temperature, allow to come to room temperature before opening to avoid cracking polyurethane hinge.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?