• USA Home
  • C4626 - Z-6-Aminohexanoic acid

EMAIL THIS PAGE TO A FRIEND
C4626 Sigma

Z-6-Aminohexanoic acid

Synonym: N-Carbobenzoxy-ε-aminocaproic acid

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis, I - Z,
InChI Key   RXQDBVWDABAAHL-UHFFFAOYSA-N
mp   54-57 °C(lit.)
storage temp.   −20°C

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?