• USA Home
  • C4714 - Calreticulin from bovine liver

EMAIL THIS PAGE TO A FRIEND
C4714 Sigma

Calreticulin from bovine liver

≥90% (SDS-PAGE), lyophilized powder

Purchase

Properties

Related Categories Calcium-associated Proteins, Cell Biology, Cell Signaling and Neuroscience, ER Resident Proteins, Endoplasmic Reticulum Stress,
assay   ≥90% (SDS-PAGE)
form   lyophilized powder
mol wt   mol wt ~60 kDa
impurities   salt, essentially free
storage temp.   −20°C
Gene Information   cow ... CALR(281036)

Description

Biochem/physiol Actions

A high-affinity calcium binding protein which plays a dynamic role in calcium homeostasis. In addition, calreticulin binds directly to the DNA domain of hormone receptors as well as to the regulatory cytoplasmic domain of proteins of the integrin family. Calreticulin has also been shown to possess a chaperone function.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

C7492 Monoclonal Anti-Calreticulin antibody produced in mouse, clone TO-11, tissue culture supernatant, buffered aqueous solution
HPA002242 Anti-CALR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000811M1 Monoclonal Anti-CALR antibody produced in mouse, clone 1G11-1A9, purified immunoglobulin, buffered aqueous solution
WH0125972M1 Monoclonal Anti-CALR3 antibody produced in mouse, clone 4E3, purified immunoglobulin, buffered aqueous solution
C4606 Anti-Calreticulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?