• USA Home
  • C4854 - Exposure cassette/BioMax® intensifying screen set
EMAIL THIS PAGE TO A FRIEND
C4854 Sigma
Exposure cassette/BioMax® intensifying screen set
for 35 × 43 cm film
Synonym: autoradiography cassettes, xray film cassettes
Purchase
Description
General description
Set contains one Kodak BioMax lightweight, vinyl-covered aluminum cassette and one Kodak BioMax MS intensifying screen. When used at ultracold temperature, allow to come to room temperature before opening to avoid cracking polyurethane hinge.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?