• USA Home
  • C4861 - ColorBurst Electrophoresis Marker

EMAIL THIS PAGE TO A FRIEND
C4861 Sigma

ColorBurst Electrophoresis Marker

High Range, mol wt 30,000-220,000 Da

Purchase

Properties

Related Categories Molecular Biology, Molecular Weight Markers, Protein Electrophoresis, Proteomics
mol wt   mol wt 30,000-220,000 Da
shipped in   wet ice
storage temp.   −20°C

Description

Components

Solution contains myosin-violet, BSA-red, GDH-blue, ADH-red, Carbonic Anhydrase-orange, 200 mM Tris, SDS and, formamide. Each lot provides the apparent molecular weights determined by comparison to an unconjugated standard.

Application

Brilliantly colored, exceptionally well-resolved, convenient, and stable, ColorBurst protein molecular weight markers perform impressively in a variety of gel compositions and concentrations. ColorBurst Markers are composed of 8 polypeptides which have been chemically reduced and conjugated to brilliantly colored dyes. Conveniently, ColorBurst Markers require no resuspension, reduction, or heating prior to use.

Colorburst can be used to estimate sample molecular weights, to monitor the progress of an electrophoretic run, or to confirm that an electroblot is complete.

Quantity

Each vial of ColorBurst contains 500 μL of solution, enough for at least 50 mini gel applications.

Legal Information

ColorBurst is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


Amicon Pro Purification System
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?