• USA Home
  • C5021 - Acetylcholinesterase from bovine erythrocytes

EMAIL THIS PAGE TO A FRIEND
C5021 Sigma

Acetylcholinesterase from bovine erythrocytes

Type XII-S, lyophilized powder, 0.25-1.0 unit/mg solid

Synonym: AChE, Acetylcholine Esterase, membrane-bound, from bovine erythrocytes, Acetylcholine acetylhydrolase

Purchase

Description

Analysis Note

The activity obtained using acetylcholine as substrate is 30-100 times that obtained with butyrylcholine, using acetylcholinesterase from electric eel.

Biochem/physiol Actions

Major degradative enzyme for acetylcholine in vivo. Converts acetylcholine + H2O to choline + acetic acid.

Other Notes

Activity believed to be membrane (stroma) bound.

Unit Definition

One unit will hydrolyze 1.0 μmole of acetylcholine to choline and acetate per min at pH 8.0 at 37 °C.

Physical form

Lyophilized powder containing phosphate buffer salts

Price and Availability

Safety & Documentation
Protocols & Articles

Articles

Cellular Derived Blood Proteins and Enzymes

Cell damage is induced by reactive oxygen species (ROS). ROS are either free radicals, reactive anions containing oxygen atoms, or molecules containing oxygen atoms that can either produce free radic...
BioFiles 2006, 1.5, 4.
Keywords: Aerobic, Alzheimer Disease, Bacterial conjugations, Biofiles, Cancer, Metabolism, Oxidations, Parkinson Disease, Reductions, Respiration

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

C3389 Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein

inhibitor

Product #

Description

Add to Cart

D7910 3,4-Dichloroisocoumarin, serine protease inhibitor
H5777 (±)-Huperzine A, synthetic, ≥98% (TLC)
A9013 1,5-Bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide
A3773 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?