• USA Home
  • C5034 - Chloroacetic acid-1-14C - unlisted item sodium salt

EMAIL THIS PAGE TO A FRIEND
C5034 Sigma

Chloroacetic acid-1-14C - unlisted item sodium salt

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2659 6.1/PG 3
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?