• USA Home
  • C5110 - Cytochrome P450 human

EMAIL THIS PAGE TO A FRIEND
C5110 Sigma

Cytochrome P450 human

4F3A isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)

Purchase

Description

Application

The CYP4F3A isoform catalyzes the 20-hydroxylation of leukotriene B4.

Biochem/physiol Actions

Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous.

Physical form

Solution in 100 mM potassium phosphate buffer, pH 7.4.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?