• USA Home
  • C5150 - Cytidine 5′-triphosphate, per­iodate oxidized sodium salt

EMAIL THIS PAGE TO A FRIEND
C5150 Sigma

Cytidine 5′-triphosphate, per­iodate oxidized sodium salt

90-95%

Synonym: Cytidine 5′-triphosphate-2′,3′-dialdehyde

  • CAS Number 75567-73-6

  • Empirical Formula (Hill Notation) C9H14N3O14P3

  • Molecular Weight 481.14

  •  MDL number MFCD00084705

  •  PubChem Substance ID 24892760

Purchase

Properties

Related Categories Aldehydes, Biochemicals and Reagents, Building Blocks, C9, Carbonyl Compounds,
InChI Key   PAUXEDHSTJAGST-POYBYMJQSA-N
assay   90-95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?