• USA Home
  • C5178 - Spectrophotometer cuvettes, silica (quartz)
EMAIL THIS PAGE TO A FRIEND
C5178 Sigma
Spectrophotometer cuvettes, silica (quartz)
Micro, with frosted wall and PTFE cover, volume ≥0.5 mL
Synonym: cuvette quartz, cuvette, cuvettes quartz
Purchase
Properties
Related Categories Labware, Spectroscopy Supplies, Spectroscopy, UV More...
material   quartz
label   A
volume   ≥0.5 mL
Description
General description
All spectrophotometer cuvettes are sold singly, not in pairs. With modern precision manufacturing methods, it is no longer necessary to pre-match cuvettes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?