• USA Home
  • C5297 - Chorionic gonadotropin human

EMAIL THIS PAGE TO A FRIEND
C5297 Sigma

Chorionic gonadotropin human

lyophilized powder, potency: ≥5,000 I.U. per mg

Synonym: Choriogonin, HCG

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chorionic Gonadotropin, Cytokines, Growth Factors and Hormones, Hormones,
sterility   sterile-filtered
form   lyophilized powder
potency   ≥5,000 I.U. per mg
mol wt   mol wt 37.9 kDa
drug control   Home Office Schedule 4.2
storage temp.   2-8°C
Gene Information   human ... CGA(1081)

Description

General description

Glycoprotein hormone consisting of a 92-amino acid α-chain which is identical to that of LH, FSH, and TSH; and a distinct 149-amino acid β-chain. Produced by chorionic tissue and responsible for maintaining the corpus luteum during early pregnancy. Also produced by trophoblast cell neoplasms, which are marked by elevated free hCG β-chains and hyperglycosylated hCG.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?