• USA Home
  • C5335 - Ethylhexadecyldimethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
C5335 Sigma

Ethylhexadecyldimethylammonium bromide

BioXtra

Synonym: Cetyldimethylethylammonium bromide, Dimethylethylhexadecylammonium bromide

  • CAS Number 124-03-8

  • Linear Formula [CH3(CH2)15N+(CH3)2CH2CH3]Br-

  • Molecular Weight 378.47

  •  Beilstein Registry Number 3729134

  •  EC Number 204-672-8

  •  MDL number MFCD00011774

  •  PubChem Substance ID 24892776

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Biochemicals and Reagents, Cationic Detergents, Detergents, Detergents A to M
description   cationic
InChI Key   VUFOSBDICLTFMS-UHFFFAOYSA-M
product line   BioXtra
composition   C16, ~85% (balance stearyl (C18) homolog)
impurities   ≤0.0005% Phosphorus (P)
  ≤0.1% Insoluble matter
ign. residue   ≤0.1%
anion traces   sulfate (SO42-): ≤0.05%
cation traces   Al: ≤0.0005%
  Ca: ≤0.0005%
  Cu: ≤0.0005%
  Fe: ≤0.0005%
  K: ≤0.005%
  Mg: ≤0.0005%
  Na: ≤0.005%
  Pb: ≤0.001%
  Zn: ≤0.0005%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BQ5425000
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?