• USA Home
  • C5356 - Greiner HLA Terasaki multiwell plates

EMAIL THIS PAGE TO A FRIEND
C5356 Sigma

Greiner HLA Terasaki multiwell plates

cover, glass

Synonym: Greiner HLA Terasaki multiwell plates

Purchase

Properties

Related Categories Antibody Plates, Antibody/Immunology Supplies, Cell Culture Supplies, Culture Multiwell Plates, Greiner,
sterility   non-sterile
feature   lid
  tissue-culture treated
mfr. no.   Greiner 653081
well size   10 μL
color   none
matrix active group   Tissue culture treated surface treatment

Description

General description

HLA Microtest plates (Terasaki) from Greiner. Designed for tissue typing techniques to ensure smooth spreading of sera and cells across the flat base of the well. The surface of each well gives a microscopically precise and clear picture. All plates are stackable.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?