• USA Home
  • C5430 - 1,2-Dimyristoyl-sn-glycerol-3-(5′-diphosphocytidine) potassium salt

EMAIL THIS PAGE TO A FRIEND
C5430 Sigma

1,2-Dimyristoyl-sn-glycerol-3-(5′-diphosphocytidine) potassium salt

~90%

Synonym: 1,2-Dimyristoyl-sn-glycero-3-diphosphocytidine

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?