• USA Home
  • C5474 - Secure-Seal hybridization chambers
EMAIL THIS PAGE TO A FRIEND
C5474 Sigma
Secure-Seal hybridization chambers
chamber L × W × D 40 mm × 22 mm × 0.8 mm
Purchase
Properties
Related Categories Genomics, PCR and Arrays, Hybridization Slide Chambers, Hybridization Supplies, Labware, Molecular Biology,
description   1 well per slide
chamber L × W × D   40 mm × 22 mm × 0.8 mm
chamber maximum volume   620 μL
Description
General description
Peel-and-stick adhesive enclosures isolate specimens on glass microscope slides. Chamber design minimizes friction, promotes reagent mixing and facilitates uniform hybridization. Sealable ports in the chamber surface allow for addition and removal of reactants. Leak-proof chambers are temperature resistant and eliminate evaporation. Secure-Seal adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating. RNase-free and ready to use.
Legal Information
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?