• USA Home
  • C5537 - (Z-Hydrazido)glycine trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
C5537 Sigma

(Z-Hydrazido)glycine trifluoroacetate salt

Synonym: Gly-NHNH-Cbz

  • CAS Number 19704-03-1

  • Empirical Formula (Hill Notation) C10H13N3O3 · C2HF3O2

  • Molecular Weight 337.25

  •  MDL number MFCD00058603

  •  PubChem Substance ID 329775092

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   ZYWUIPUGRJNRCM-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?