| Related Categories | Conductivity Meters, Hanna, Hanna Conductivity, Labware, Manufacturer Browser More... |
| feature | temperature 10 ‑ 40 °C with built-in temperature sensor; operating range 0-50°C; adjustable temperature coefficient 0-2.5% per °C |
| measuring range | 0-1999 mS/cm conductivity, accuracy: 1% full scale |
| size | 164 mm × 76 mm × 45 mm |
| battery power | 1 × 9 V |
HI 8733 is a splashproof multi-range range meter that measures conductivity in four different ranges for the highest accuracy possible. Measurements in each of the four rnages can be performed at the touch of a button without having to change the probe. This allows the user to easily switch applications without having to re-calibrate. Simple manual temperature compensation is acheived through a knob on the front panel.
Supplied complete with HI 76302W probe with internal temperature sensor and 1 M cable, calibration screwdriver, calibration solutions, protective case, battery and instructions.
If the temperature of the solution fluctuates, HI8733 with a built in temperature sensor and automatic temperature compensation is the right choice.