• USA Home
  • C5588 - Anti-Cyclin D1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
C5588 Sigma

Anti-Cyclin D1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Proteins/Bioactives/Markers/Receptors for Stem Cell Biology, Antibodies for Cell Biology, Antibodies for Cell Cycle,
species reactivity   human, rat
application(s)   microarray: suitable
  western blot: 10 μg/mL using human and rat cell extracts
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 34 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CCND1(595)
rat ... Ccnd1(58919)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

recombinant human Cyclin D1

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Specificity

No cross-reactivity is detected with other cyclins (Cyclin A, B or E).

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?