• USA Home
  • C5646 - Cardiolipin solution from bovine heart

EMAIL THIS PAGE TO A FRIEND
C5646 Sigma

Cardiolipin solution from bovine heart

~5 mg/mL in methanol, ≥97% (TLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophosphoglycerophosphoglycerols (Cardiolipins), Glycerophospholipid Metabolism, Glycerophospholipids,
assay   ≥97% (TLC)
concentration   ~5 mg/mL in methanol
storage temp.   −20°C

Description

Analysis Note

Actual concentration given on label.

Biochem/physiol Actions

Cardiolipin is a mitochondrial phospholipid that is found in mammalian tissues in low concentrations. It is often a membrane component. Inhibits cell attachment of fibronectin, vitronectin and Type I collagen.

Other Notes

>80% polyunsaturated fatty acid content, primarily linoleic acid.

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

[JIM BALL TEST DOC] MIDA-protected Boronate Esters

The Suzuki-Miyaura cross-coupling reaction is one of the most important and highly utilized reactions in organic chemistry, with applications in polymer science as well as in the fine chemicals and p...
Josephine Nakhla
ChemFiles Volume 9 Article 1
Keywords: Aminations, Applications, Bromoborations, Building blocks, Catalysis, Chemfiles, Chromatography, Coupling reactions, Cross couplings, Cross metathesis, Cyclopropanations, Epoxidations, Industries, Ligands, Metathesis, Methods, Natural product synthesis, Olefin metathesis, Pharmaceutical, Polymer science, Protocols, Purification, Reductions, Reductive aminations, Stille coupling, Suzuki coupling, Suzuki reactions, Takai olefination, Transmetalation, Type, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?