• USA Home
  • C5845 - Corn meal peptone yeast agar

EMAIL THIS PAGE TO A FRIEND
C5845 Sigma

Corn meal peptone yeast agar

for microbiology

Purchase

Properties

grade   for microbiology
pH   6.5±0.2 (25 °C)

Description

Components

Components (g/L)
Peptic Digest of Animal Tissue 10.00
Corn Meal 20.00
Dextrose 10.00
Yeast Extract 4.00
Agar 20.00

Application

Recommended for chlamydospore production by Candida albicans and the maintenance of fungal stock cultures.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?