EMAIL THIS PAGE TO A FRIEND
C5879 Sigma

Cinnamalfluorene

Synonym: Cinnamylidenefluorene

  • CAS Number 2871-26-3

  • Empirical Formula (Hill Notation) C22H16

  • Molecular Weight 280.36

  •  MDL number MFCD00019099

  •  PubChem Substance ID 24892814

Purchase

Properties

InChI Key   FDELSKWIDIUQDN-DHZHZOJOSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?