• USA Home
  • C5881 - Hexadecylpyridinium bromide

EMAIL THIS PAGE TO A FRIEND
C5881 Sigma

Hexadecylpyridinium bromide

Synonym: Cetylpyridinium bromide

  • CAS Number 140-72-7

  • Empirical Formula (Hill Notation) C21H38BrN

  • Molecular Weight 384.44

  •  Beilstein Registry Number 3778948

  •  EC Number 205-428-3

  •  MDL number MFCD00011988

  •  PubChem Substance ID 24892816

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
UU4848000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?