• USA Home
  • C5921 - Cholesterol Esterase from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
C5921 Sigma

Cholesterol Esterase from bovine pancreas

lyophilized powder, ≥10,000 units/g protein

Synonym: Sterol-ester acylhydrolase

Purchase

Description

Analysis Note

Protein determined by biuret.

Unit Definition

One unit will hydrolyze 1.0 μmole of cholesteryl oleate to cholesterol and oleic acid per min at pH 7.0 at 37 °C in the presence of taurocholate.

Application

Enzyme responsible for the hydrolysis of many of the fatty acid esters of cholesterol.
Optimum pH range: 6-8
Activators: Bisphenol A diglycidyl ether, cAMP-dependent protein kinase type II, ethanol, methanol, n-butanol, phosphatidylcholine, phosphatidylethanolamine, sodium taurocholic acid
Inhibitors: Bisphenol A methacrylate, diisopropylfluorophosphate, enolase, Hg2+, sodium fluoride, phosphatidic acid, phosphatidylcholine, phosphatidylserine

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?