• USA Home
  • C5937 - Nalgene® sealing cap assembly
EMAIL THIS PAGE TO A FRIEND
C5937 Sigma
Nalgene® sealing cap assembly
For T 2168
Purchase
Properties
Related Categories 50 mL centrifuge tubes, Centrifugation Supplies, Centrifuge Tubes, Plastic, Labware, Manufacturer Browser,
material   Tefzel® plug
  Viton® O-ring
  cap screw top
  polypropylene screw closure
packaging   pack of 2 ea
mfr. no.   Nalgene
Description
Application
Use with recommended tubes and bottles for leakproof service at maximum speed. Also recommended for spinning hazardous materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?