• USA Home
  • C6105 - Cellobiase from Aspergillus niger

EMAIL THIS PAGE TO A FRIEND
C6105 Sigma

Cellobiase from Aspergillus niger Green Alternative

liquid, ≥250 units/g

Synonym: Novozyme 188

Purchase

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

General description

Cellobiase enzyme preparation obtained by submerged fermentation of an Aspergillus niger microorganism. The cellobiase hydrolyzes cellobiose to glucose.

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles.

Legal Information

A product of Novozyme Corp.

Application

Cellobiase from Aspergillus niger has been used in a study to assess the targeted antifungal delivery system of a β-Glucosidase sensitive nystatin–star poly(ethylene glycol) conjugate. Cellobiase from Aspergillus niger has also been used in a study to investigate the influence of substrate particle size and wet oxidation on physical surface structures and enzymatic hydrolysis of wheat straw.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
What is the unit definition of Cellobiase, Product C6105?
One unit of cellobiase activity (CBU) is defined as 2 μmole of glucose produced per minute at 40°C, pH 5.
What is the density of Cellobiase, Product C6105?
The density of this enzyme solution is approximately 1.2 g/mL.
What is the recommended dosage for Cellobiase, Product C6105?
The manufacturer's recommended dosage is 0-5%. That is, the enzyme solution should be be used at an enzyme:substrate (w:w) ratio of at least 1:20.
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
Show more questions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?