EMAIL THIS PAGE TO A FRIEND
C6110 Sigma

Crebtide

≥98% (HPLC), lyophilized powder

  • Empirical Formula (Hill Notation) C79H141N31O20

  • Molecular Weight 1845.16

  •  MDL number MFCD03453644

  •  PubChem Substance ID 329775113

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology, Protein Kinase Related Peptides,
InChI Key   KCZQVMUDQWHYKI-ZPXCENDESA-N
assay   ≥98% (HPLC)
form   lyophilized powder
composition   Peptide content, ~80%
solubility   H2O: soluble
storage temp.   −20°C

Description

Amino Acid Sequence

Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg-Lys

Other Notes

Peptide corresponding to the consensus phosphorylation site at Ser133 of the cyclic AMP response element binding protein (CREB).

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?