• USA Home
  • C6213 - Cos-1 cells from African green monkey kidney

EMAIL THIS PAGE TO A FRIEND
C6213 Sigma

Cos-1 cells from African green monkey kidney

Synonym: ECACC 88031701

Purchase

Properties

shipped in   dry ice
storage temp.   −70°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?