• USA Home
  • C6312 - Nalgene® sealing cap assembly
EMAIL THIS PAGE TO A FRIEND
C6312 Sigma
Nalgene® sealing cap assembly
Purchase
Properties
Related Categories Centrifugation Supplies, Centrifuge Tubes, Plastic, Centrifuge accessories, Labware, Manufacturer Browser,
material   Tefzel® plug
  Viton® O-ring
  polypropylene screw closure
mfr. no.   Nalgene
Description
Application
Use with recommended tubes and bottles for leakproof service at maximum speed. Also recommended for spinning hazardous materials.
General description
Polypropylene shoulder support ring prevents bending or collapse of neck.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Steriflip Filters Units
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?