• USA Home
  • C6354 - Anti-CINC-1 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
C6354 Sigma

Anti-CINC-1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-Cytokine-Induced Neutrophil Chemoattractant-1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Chemokines, Antibodies to Cytokines and Growth Factors,
species reactivity   rat
application(s)   immunohistochemistry: 5-15 μg/mL
  neutralization: suitable
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... Cxcl1(81503)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant rat CINC-1, expressed in E. coli.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Specificity

The antibody has the ability to neutralize the biological activity of recombinant rat CINC-1. It will not neutralize the activity of recombinant human GROα, GROβ, or GROγ. Anti-CINC-1 exhibits less than 50% cross-reactivity with recombinant mouse KC and less than 10% cross-reactivity with recombinant human GROα, GROβ, GROγ, and mouse MIP-2.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?