EMAIL THIS PAGE TO A FRIEND
C6389 Sigma

Chromosorb® W/AW

45-60 mesh

Synonym: Diatomaceous earth, flux-calcined

Purchase

Properties

InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
particle size   45-60 mesh
surface area   1.0 m2/g

Description

Preparation Note

Acid Washed

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?