• USA Home
  • C6393 - Carboxypeptidase A Type I-PMSF from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
C6393 Sigma

Carboxypeptidase A Type I-PMSF from bovine pancreas

~50 units/mg protein

Purchase

Properties

storage temp.   2-8°C

Description

Analysis Note

Protein determined by E1%/278

Preparation Note

C 0261 treated with phenylmethylsulfonyl fluoride to eliminate trypsin and chymotrypsin activity. Dialyzed and recrystallized: aqueous suspension with toluene added.

Unit Definition

One unit will hydrolyze 1.0 μmole of hippuryl-L-phenylalanine per min at pH 7.5 at 25°C, unless otherwise indicated.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

C9268 Carboxypeptidase A from bovine pancreas, (Type II-PMSF treated), ≥50 units/mg protein, aqueous suspension

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?