• USA Home
  • C6418 - Combs for horizontal submarine electrophoresis unit
EMAIL THIS PAGE TO A FRIEND
C6418 Sigma
Combs for horizontal submarine electrophoresis unit
size 1.0 mm × 16 wells, for Mini-gel 7.5 cm wide
Purchase
Description
Features and Benefits
Horizontal Gel Comb 1.0 mm x 16 wells, for Mini-gel 7.5 cm wide Horizontal units. Precision-machined polycarbonate. With adjustable backing, can be adjusted to the desired height above the gel tray. Designed for multi-channel pipettor loading.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?