• USA Home
  • C6426 - Tyr-Corticotropin Releasing Factor Human, Rat

EMAIL THIS PAGE TO A FRIEND
C6426 Sigma

Tyr-Corticotropin Releasing Factor Human, Rat

≥97% (HPLC)

  • CAS Number 100513-58-4

  • Empirical Formula (Hill Notation) C217H353N61O65S2

  • Molecular Weight 4920.62

  •  MDL number MFCD00081236

Purchase

Properties

Related Categories CRF, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology,
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... CRH(1392)

Description

Amino Acid Sequence

Tyr-Ser-Glu-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-Leu-Met-Glu-Ile-Ile-NH2

Application

CRF analog used to prepare iodinated CRF for receptor studies and immunoassay

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?