• USA Home
  • C6430 - N-Cbz-Ala-Ala-Phe isoluminol

EMAIL THIS PAGE TO A FRIEND
C6430 Sigma

N-Cbz-Ala-Ala-Phe isoluminol

≥98% (HPLC)

Purchase

Properties

assay   ≥98% (HPLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?