• USA Home
  • C6432 - Nunc® Lab-Tek® Chamber Slide system
EMAIL THIS PAGE TO A FRIEND
C6432 Sigma
Nunc® Lab-Tek® Chamber Slide system
1 well on Permanox®
Synonym: chamber slides, microscope slides, microscopy slides
Purchase
Properties
Related Categories Cell Chamber Slides and Glass Coverslips, Cell Culture Supplies, Lab-Tek Chamber Slide system, Lab-Tek® Chamber Slides, Labware,
sterility   sterile
feature   CE marked
packaging   pack of 16
  case of 96
mfr. no.   Nunc 177410
suitability   suitable for (viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology)
Description
General description
Nunc® Lab-Tek® Chamber Slide System
• Especially developed for microscopy of in situ tissue culture
• Cells grow on a standard microscope slide
• Growth chamber can be removed when culturing is complete
• Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
• Broad range of formats and well numbers
• Fits standard equipment
• Certified surface treatment ensures optimal contitions for cell attachment and growth
• Saves time and reagents
• Permanox® does not interfere with fluorescent labels
• No cell transfer needed prior to visualisation/staining
• Upper structure can be removed when culturing is complete
• Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
• Broad range of formats and well numbers
• Suitable for use with fluorescent labels
• CE marked
Legal Information
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Permanox is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?