• USA Home
  • C6447 - Chorionic gonadotropin α subunit human

EMAIL THIS PAGE TO A FRIEND
C6447 Sigma

Chorionic gonadotropin α subunit human

recombinant, expressed in mouse cell line, powder

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chorionic Gonadotropin, Cytokines, Growth Factors and Hormones, Hormones,
recombinant   expressed in mouse cell line
form   powder
storage temp.   −20°C
Gene Information   human ... CGB5(93659)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?