• USA Home
  • C6479 - Anti-CINC-2α antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
C6479 Sigma

Anti-CINC-2α antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-Cytokine-Induced Neutrophil Chemoattractant-2α

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Chemokines, Antibodies to Cytokines and Growth Factors,
species reactivity   rat
application(s)   indirect ELISA: 0.5-1.0 μg/mL
  neutralization: suitable
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... Cxcl3(171551)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant rat CINC-2α, expressed in E. coli.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Specificity

The antibody recognizes recombinant rat CINC-2α. It exhibits greater than 50% cross-reactivity with recombinant rat CINC-1, rat CINC-2β, mouse KC, mouse MIP-2 and less than 25% cross-reactivity with recombinant human IL-8, GROα, GROβ, and GROγ.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?