EMAIL THIS PAGE TO A FRIEND
C6501 Sigma

L-Cysteine(Z)-OH

  • CAS Number 1625-72-5

  • Empirical Formula (Hill Notation) C11H13NO4S

  • Molecular Weight 255.29

  •  MDL number MFCD00055774

  •  PubChem Substance ID 24892871

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   IAZGSEXFFGHKDF-VIFPVBQESA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?