• USA Home
  • C6545 - NG,NG,-Di-Cbz-L-arginine

EMAIL THIS PAGE TO A FRIEND
C6545 Sigma

NG,NG,-Di-Cbz-L-arginine

  • CAS Number 4125-79-5

  • Empirical Formula (Hill Notation) C22H26N4O6

  • Molecular Weight 442.47

  •  MDL number MFCD00083284

  •  PubChem Substance ID 24892878

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   YPUMRXRWHYQIPH-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?