• USA Home
  • C6570 - Petri-Pad culture dish system

EMAIL THIS PAGE TO A FRIEND
C6570 Sigma

Petri-Pad culture dish system

culture dishes only

Synonym: dish culture, petri culture, petri dishes plastic

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Dishes, Labware, Microbiology Equipment, Petri Dishes More...
material   polystyrene
  round
sterility   sterile
packaging   box of 100 Millipore Petri Dish (Single use)
mfr. no.   Millipore, PD1004700
H × diam.   9 mm × 47 mm
matrix active group   not treated surface treatment

Description

General description

A complete system for growing microorganisms on a "solid" surface with liquid broth media. Just add sterile broth to saturate the pure absorbent cellulose pad, and the dish is ready. Dishes without pads can be used for agar-based solid media.
The dishes are optically clear polystyrene with lips on both the base and the cover for easy one-hand manipulation. There is a frosted area on the bottom for marking. The 47 mm diameter allows culturing on standard-size filter membranes.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?