• USA Home
  • C6610 - Culturewell coverslip system
EMAIL THIS PAGE TO A FRIEND
C6610 Sigma
Culturewell coverslip system
2 wells, diam. × D 15 mm × 1.0 mm, sterile
Synonym: coverslips, microscope slide, microscope slides
Purchase
Properties
Related Categories Cell Chamber Slides and Glass Coverslips, Cell Culture Supplies, Labware More...
sterility   sterile
mfr. no.   Grace Bio-Labs
diam. × D   15 mm × 1.0 mm
Description
General description
Culturewell Coverslip System − Silicone gaskets secured to 24 × 50 mm, no. 11/2 glass coverslips. 9 mm spacing for multichannel pipettors. Leak-proof enclosures eliminate well-to-well cross contamination. Design provides convenient coverslip handling using microscope slide holders and accessories. Gaskets remove cleanly, easily for confocal microscopy and high throughput screening. Available as inserts for batch processing and as individual coverslips. Chambered Coverslip Inserts. Each package contains 10 sterile pouches, each containing 4 coverslips, and 10 sterile culture plates.
Legal Information
Culturewell is a trademark of Grace Bio-Labs, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?