• USA Home
  • C6638 - Creatine Phosphokinase from rabbit brain

EMAIL THIS PAGE TO A FRIEND
C6638 Sigma

Creatine Phosphokinase from rabbit brain

Type IV, lyophilized powder, 50-150 units/mg protein

Synonym: ATP: Creatine N-Phosphotransferase, CPK, Creatine Kinase, Phosphocreatine phosphokinase

Purchase

Description

Analysis Note

Protein determined by biuret.

Biochem/physiol Actions

Phosphorylates ADP to form ATP using phosphocreatine as the phosphate donor.

Caution

The use of 0.1% albumin in the reaction buffer is recommended to avoid inactivation due to dilution.

Unit Definition

One unit will transfer 1.0 μmole of phosphate from phosphocreatine to ADP per min at pH 7.4 at 30 °C.

Physical form

Lyophilized powder containing buffer salt as glycine, pH 9.0.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

inhibitor

Product #

Description

Add to Cart

C5913 4-Chloromercuribenzoic acid, cysteine active site modifier

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?