EMAIL THIS PAGE TO A FRIEND
C6640 Sigma

8-Chlorotheophylline

Synonym: 8-Chloro-1,3-dimethyl-2,6(1H,3H)-purinedione

Purchase

Properties

mp   290 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
XH5063000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?