• USA Home
  • C6695 - Petri-Pad culture dish system

EMAIL THIS PAGE TO A FRIEND
C6695 Sigma

Petri-Pad culture dish system

Complete unit

Synonym: dish culture, petri culture, petri dishes plastic

Purchase

Properties

Related Categories Cell Culture Supplies, Culture Dishes, Labware, Microbiology Equipment, Petri Dishes More...
material   polystyrene
  round
description   Unit consists of a 47 mm culture dish and one Millipore pure cellulose Petri-Pad
sterility   sterile
packaging   sleeve of 20 (30 x 20 sleeves, or 600 total)
mfr. no.   Millipore, PD20047S5
  Millipore, PD20047S5
H × diam.   9 mm × 47 mm
matrix active group   not treated surface treatment

Description

General description

A complete system for growing microorganisms on a "solid" surface with liquid broth media. Just add sterile broth to saturate the pure absorbent cellulose pad, and the dish is ready. Dishes without pads can be used for agar-based solid media.
The dishes are optically clear polystyrene with lips on both the base and the cover for easy one-hand manipulation. There is a frosted area on the bottom for marking. The 47 mm diameter allows culturing on standard-size filter membranes.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?