• USA Home
  • C6723 - Monoclonal Anti-CXCR5 (BLR-1) antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
C6723 Sigma

Monoclonal Anti-CXCR5 (BLR-1) antibody produced in mouse

clone 51505.111, purified immunoglobulin, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Chemokine Receptors, Antibodies to Cytokines and Growth Factors,
species reactivity   human
application(s)   flow cytometry: suitable
clone   51505.111, monoclonal
antibody form   purified immunoglobulin
form   lyophilized powder
isotype   IgG2b
storage temp.   −20°C
Gene Information   human ... CXCR5(643)
mouse ... Cxcr5(12145)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

human CXCR5 transfected NSO mouse myeloma cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Specificity

The antibody reacts with human CXCR5 transfectant cells and not the parent cell line. The antibody shows no cross-reactivity with CXCR2, CXCR3, or CXCR4.

Preparation Note

Purified using protein G.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?