• USA Home
  • C6742 - Carboxypeptidase W from wheat

EMAIL THIS PAGE TO A FRIEND
C6742 Sigma

Carboxypeptidase W from wheat

≥50 units/mg protein

Purchase

Description

Physical form

One unit will hydrolyze 1.0 μmole of N-CBZ-L-Phe-L-Ala to N-CBZ-L-Phe and L-Ala per minute at pH 4.0 at 30 °C

Suplied as a suspension in sodium acetate with 2.5 M NaCl, pH 4.0

Application

Carboxypeptidase W from wheat has been used in a study to assess the proteolytic activities in dormant rye (Secale cereale L.) grain. Carboxypeptidase W from wheat has also been used in a study to investigate the structure determination of the human protective protein.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?