• USA Home
  • C6760 - Cytosine β-D-arabinofuranoside 5′-monophosphate

EMAIL THIS PAGE TO A FRIEND
C6760 Sigma

Cytosine β-D-arabinofuranoside 5′-monophosphate

≥99%

Synonym: Ara-CMP

  • CAS Number 7075-11-8

  • Empirical Formula (Hill Notation) C9H14N3O8P

  • Molecular Weight 323.20

  •  MDL number MFCD00069771

  •  PubChem Substance ID 24892888

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
InChI Key   IERHLVCPSMICTF-CCXZUQQUSA-N
assay   ≥99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?