• USA Home
  • C6828 - Carboxanine fd step IV

EMAIL THIS PAGE TO A FRIEND
C6828 Sigma

Carboxanine fd step IV

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

not to be listed in catalog

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?