EMAIL THIS PAGE TO A FRIEND
C6859 Sigma

Cefprozil

solid

Purchase

Properties

form   solid
mol wt   apparent mol wt 407.45

Description

Application

Antibacterial

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XI0339000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?